This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04335709_m1
See other SMPX GE Assays ›
Gene Symbol:
Smpx
Gene Name:
small muscle protein, X-linked
Gene Aliases:
1010001C09Rik, Csl
Chromosome Location:
Chr.X: 157698973 - 157752591 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04335709_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Smpx
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_045569.1 3 - 4 443 87
GenBank mRNA W18646.1 - 3 - 4 156 87

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream membrane-bound transcription factor peptidase,
site 2
Mbtps2 9630032G22Rik,
AI662535
-108.2kb Anti-sense Chr. X - 157547822 - 157598715 on NCBI Build GRCm38
Downstream predicted gene 15169 Gm15169 OTTMUSG00000019393 42.5kb Anti-sense Chr. X - 157757180 - 157759273 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0X00001601.mm.1

Set Membership:

Amplicon length between 86 and 100 Mus musculus Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0X00010065.mm.1 NR_045569.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00010065.mm.1 NR_045569.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00010065.mm.1 TC0X00001601.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0