This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04335499_m1
See other 4922502N22RIK GE Assays ›
Gene Symbol:
4922502N22Rik
Gene Name:
RIKEN cDNA 4922502N22 gene
Gene Aliases:
-
Chromosome Location:
Chr.6: 139315597 - 139322576 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04335499_m1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 4922502N22Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_045149.1 1 - 2 71 79
GenBank mRNA AK015023.1 - 1 - 2 72 79
AK132719.1 - 1 - 2 72 79

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream immunoglobulin (CD79A) binding protein 1b Igbp1b alpha4-b -662.4kb Sense Chr. 6 - 138657089 - 138658444 on NCBI Build GRCm38
Downstream RERG/RAS-like Rergl EG632971 170.6kb Sense Chr. 6 - 139493182 - 139501909 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600003349.mm.1
This assay targets the same location as RTA transcript cluster(s) TC0400003997.rn

Set Membership:

Amplicon length between 71 and 85 Mus musculus Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600026337.mm.1 NR_045149.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600026337.mm.1 NR_045149.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600026337.mm.1 TC0600003349.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600026338.mm.1 NR_045149.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600026338.mm.1 NR_045149.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600026338.mm.1 TC0600003349.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0