This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04335476_s1
See other MIR5128 GE Assays ›
Gene Symbol:
Mir5128
Gene Name:
microRNA 5128
Gene Aliases:
mir-5128, mmu-mir-5128, mu-mir-5128
Chromosome Location:
Chr.2: 37688201 - 37688284 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04335476_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mir5128
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_039590.1 1 46 64
GenBank mRNA JF446431.1 - 1 46 64

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream RAB GTPase activating protein 1 Rabgap1 Gapcena,
mKIAA4104
-121.7kb Anti-sense Chr. 2 - 37443261 - 37566454 on NCBI Build GRCm38
Downstream crumbs family member 2 Crb2 5930402A21,
BC043114
87.9kb Anti-sense Chr. 2 - 37776225 - 37799103 on NCBI Build GRCm38

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-c694fbbc8-74xkl:80/100.66.128.6:80.
git-commit: 2cf2d5f40d96ab2bfa499d940143ca22e465fdb8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0