This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04335466_s1
See other MIR5107 GE Assays ›
Gene Symbol:
Mir5107
Gene Name:
microRNA 5107
Gene Aliases:
mir-5107, mmu-mir-5107, mu-mir-5107
Chromosome Location:
Chr.18: 60812076 - 60812163 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04335466_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mir5107
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_039567.1 1 42 64
GenBank mRNA JF446407.1 - 1 42 64
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA CR522355.1 - 2 Chr. 18 - 60812001 - 60812652 [-] on Build GRCm38 64

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream ribosomal protein S14 Rps14 2600014J02Rik,
AL023078
-33.5kb Sense Chr. 18 - 60774596 - 60778546 on NCBI Build GRCm38
Downstream Treacher Collins Franceschetti syndrome 1,
homolog
Tcof1 AA408847,
AW209012,
treacle
1.6kb Anti-sense Chr. 18 - 60813736 - 60848983 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1800004415.mm.1 NR_039567.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800004415.mm.1 NR_039567.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800004415.mm.1 TC1800000617.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0