This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04335130_m1
See other 1700012D14RIK GE Assays ›
Gene Symbol:
1700012D14Rik
Gene Name:
RIKEN cDNA 1700012D14 gene
Gene Aliases:
-
Chromosome Location:
Chr.7: 111117673 - 111122675 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04335130_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 1700012D14Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_015573.2 2 - 3 408 111
GenBank mRNA AK005898.1 - 2 - 3 410 111
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY706038.1 - 2 - 3 409 Chr. 7 - 111117673 - 111122675 [-] on Build GRCm38 111
GenBankmRNA AV040400.2 - 1 - 2 102 Chr. 7 - 111117673 - 111121010 [-] on Build GRCm38 111
GenBankmRNA BE633713.1 - 2 - 3 290 Chr. 7 - 111117746 - 111122682 [-] on Build GRCm38 111

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream eukaryotic translation initiation factor 4,
gamma 2
Eif4g2 AA589388,
DAP-5,
E130105L11Rik,
Nat1,
Natm1,
p97
-34.6kb Sense Chr. 7 - 111067985 - 111083030 on NCBI Build GRCm38
Downstream UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 18 Galnt18 2900011G21Rik,
BC024988,
Galntl4
350.6kb Sense Chr. 7 - 111471656 - 111780067 on NCBI Build GRCm38

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0