This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04335099_g1
See other A230056P14RIK GE Assays ›
Gene Symbol:
A230056P14Rik
Gene Name:
RIKEN cDNA A230056P14 gene
Gene Aliases:
OTTMUSG00000027345
Chromosome Location:
Chr.7: 55962531 - 55980096 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04335099_g1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: A230056P14Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_015495.2 4 - 5 520 111
GenBank mRNA AK043630.1 - 4 - 5 521 111
AK140288.1 - 4 - 5 521 111
BC147226.1 - 2 - 3 159 111
BC147227.1 - 2 - 3 159 111
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BB641255.1 - 4 - 5 511 Chr. 7 - 55962540 - 55976943 [+] on Build GRCm38 111
GenBankmRNA BY727408.1 - 4 - 5 510 Chr. 7 - 55962541 - 55976908 [+] on Build GRCm38 111

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream non imprinted in Prader-Willi/Angelman syndrome 2 homolog (human) Nipa2 2600017P10Rik,
3830408P04Rik,
AB041581
-13.9kb Anti-sense Chr. 7 - 55931266 - 55962655 on NCBI Build GRCm38
Downstream non imprinted in Prader-Willi/Angelman syndrome 1 homolog (human) Nipa1 1110027G09Rik,
A830014A18Rik,
FSP3,
Spg6
1.5kb Anti-sense Chr. 7 - 55978484 - 56019573 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK140288.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700008497.mm.1 NR_015495.2 Refseq
Affymetrix Mouse Clariom S Array PSR0700008497.mm.1 TC0700000934.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0