This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04278842_m1
See other 4930558G05RIK GE Assays ›
Gene Symbol:
4930558G05Rik
Gene Name:
RIKEN cDNA 4930558G05 gene
Gene Aliases:
-
Chromosome Location:
Chr.X: 129051524 - 129067146 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04278842_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 4930558G05Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_045441.1 2 - 3 218 148
GenBank mRNA AK016174.1 - 2 - 3 221 148
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY715778.1 - 2 - 3 221 Chr. X - 129051524 - 129067146 [+] on Build GRCm38 148

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream predicted gene 4916 Gm4916 EG237030,
Rbm31x
-687.1kb Sense Chr. X - 128375395 - 128377169 on NCBI Build GRCm38
Downstream diaphanous homolog 2 (Drosophila) Diap2 Dia3,
Diaph2,
Drf2,
E430022I22Rik
683.3kb Sense Chr. X - 129749668 - 130465834 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0X00001252.mm.1

Set Membership:

Amplicon length greater than or equal to 101 Mus musculus Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0X00007750.mm.1 NR_045441.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00007750.mm.1 NR_045441.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00007750.mm.1 TC0X00001252.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0X00007747.mm.1 NR_045441.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00007747.mm.1 NR_045441.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00007747.mm.1 TC0X00001252.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0X00007749.mm.1 NR_045441.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00007749.mm.1 NR_045441.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00007749.mm.1 TC0X00001252.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0