This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04278677_m1
See other A430088P11RIK GE Assays ›
Gene Symbol:
A430088P11Rik
Gene Name:
RIKEN cDNA A430088P11 gene
Gene Aliases:
-
Chromosome Location:
Chr.15: 80691025 - 80697610 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04278677_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: A430088P11Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_045310.1 2 - 3 439 90
GenBank mRNA AK040361.1 - 2 - 3 439 90

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream family with sequence similarity 83,
member F
Fam83f AW544981 Anti-sense Chr. 15 - 80671847 - 80700425 on NCBI Build GRCm38
Downstream trinucleotide repeat containing 6b Tnrc6b 2700090M07Rik,
A730065C02Rik,
AI848765,
D230019K20Rik
14.3kb Anti-sense Chr. 15 - 80711313 - 80941086 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500001909.mm.1

Set Membership:

Amplicon length between 86 and 100 3' Most Mus musculus Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500014990.mm.1 NR_045310.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500014990.mm.1 NR_045310.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500014990.mm.1 TC1500001909.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0