This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04278590_m1
See other 4932702P03RIK GE Assays ›
Gene Symbol:
4932702P03Rik
Gene Name:
RIKEN cDNA 4932702P03 gene
Gene Aliases:
1700047I16Rik
Chromosome Location:
Chr.13: 24905600 - 24911780 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04278590_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 4932702P03Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_045182.1 1 - 2 105 104
GenBank mRNA CJ096614.1 - 1 - 2 85 104

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream aldhehyde dehydrogenase family 5,
subfamily A1
Aldh5a1 6330403E24Rik,
Ahd-1,
Ahd1,
D630032B01Rik,
OTTMUSG00000000613,
SSADH,
Ssdh1
Anti-sense Chr. 13 - 24907579 - 24937661 on NCBI Build GRCm38
Downstream cDNA RIKEN 9330162012 gene 9330162012Rik 26.6kb Sense Chr. 13 - 24937401 - 24939049 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1300000282.mm.1

Set Membership:

Amplicon length greater than or equal to 101 3' Most Mus musculus Probe spans exons

MicroArray Probes:

Agilent Affymetrix

Array Name Probe IDs Targets Target Type
Whole Mouse Genome G4122A A_52_P440119 AK006712.1 mouse_transcriptome
Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1453353_PM_at - -
Mouse Genome 430 2.0 Array 1453353_PM_at - -
Mouse Genome 430 2.0 Array 1453353_at - -
Affymetrix Mouse Clariom S Array PSR1300001721.mm.1 NR_045182.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300001721.mm.1 NR_045182.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300001721.mm.1 TC1300000282.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300001724.mm.1 NR_045182.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300001724.mm.1 NR_045182.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300001724.mm.1 TC1300000282.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300001725.mm.1 NR_045182.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300001725.mm.1 NR_045182.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300001725.mm.1 TC1300000282.mm.1 MTA TC
Mouse Expression Set 430 1453353_at - -
Mouse Expression Set 430 1453353_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1453353_at - -
Affymetrix HT Mouse Genome 430B Array 1453353_PM_at - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0