This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04278408_m1
See other 4930480G23RIK GE Assays ›
Gene Symbol:
4930480G23Rik
Gene Name:
RIKEN cDNA 4930480G23 gene
Gene Aliases:
-
Chromosome Location:
Chr.4: 19979445 - 20005739 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04278408_m1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 4930480G23Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_040768.1 4 - 5 880 149
GenBank mRNA AK015599.1 - 4 - 5 880 149

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream ATPase,
H+ transporting,
lysosomal V0 subunit D2
Atp6v0d2 1620401A02Rik,
AI324824,
V-ATPase
-56.9kb Sense Chr. 4 - 19876838 - 19922566 on NCBI Build GRCm38
Downstream tocopherol (alpha) transfer protein Ttpa alpha-TTP 24.5kb Anti-sense Chr. 4 - 20008089 - 20030785 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400002277.mm.1

Set Membership:

Amplicon length greater than or equal to 101 3' Most Mus musculus Probe spans exons

MicroArray Probes:

Agilent Affymetrix

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2361927 AK015599.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 219824 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 495177 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 591217 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 163774 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 714502 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 731439 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 621658 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 678101 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 643129 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 208562 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 947547 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 460173 AK078258.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 460173 AY129229.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 460173 BC028808.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 460173 BC131949.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 460173 NM_020492.3 refseq_mrna_mouse_NM
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 460173 XM_001472063.1 refseq_mrna_mouse_XM
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 45340 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 130121 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2361927 AK015599.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P137479 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22767611 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22767672 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22767603 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22767677 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22767612 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22767682 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22767679 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22767600 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22767676 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22767673 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22767615 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22767601 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22767617 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22767675 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22767616 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22767609 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22767671 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22767607 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22767605 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22767604 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22767670 - -
Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1453707_PM_at - -
Mouse Genome 430 2.0 Array 1453707_PM_at - -
Mouse Genome 430 2.0 Array 1453707_at - -
Affymetrix Mouse Clariom S Array PSR0400018888.mm.1 NR_040768.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400018888.mm.1 NR_040768.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400018888.mm.1 TC0400002277.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400018889.mm.1 NR_040768.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400018889.mm.1 NR_040768.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400018889.mm.1 TC0400002277.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400018891.mm.1 NR_040768.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400018891.mm.1 NR_040768.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400018891.mm.1 TC0400002277.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400018895.mm.1 NR_040768.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400018895.mm.1 NR_040768.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400018895.mm.1 TC0400002277.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400018894.mm.1 NR_040768.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400018894.mm.1 NR_040768.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400018894.mm.1 TC0400002277.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400018890.mm.1 NR_040768.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400018890.mm.1 NR_040768.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400018890.mm.1 TC0400002277.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400018893.mm.1 NR_040768.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400018893.mm.1 NR_040768.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400018893.mm.1 TC0400002277.mm.1 MTA TC
Mouse Expression Set 430 1453707_PM_at - -
Mouse Expression Set 430 1453707_at - -
Affymetrix HT Mouse Genome 430B Array 1453707_at - -
Affymetrix HT Mouse Genome 430B Array 1453707_PM_at - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header