This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04278294_m1
See other A730056A06RIK GE Assays ›
Gene Symbol:
A730056A06Rik
Gene Name:
RIKEN cDNA A730056A06 gene
Gene Aliases:
C76391
Chromosome Location:
Chr.7: 73313806 - 73375774 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04278294_m1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: A730056A06Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_040325.1 1 - 2 253 104
GenBank mRNA AK043101.1 - 1 - 2 245 104
AK053954.1 - 1 - 2 219 104
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BB665695.1 - 1 - 2 219 Chr. 7 - 73316000 - 73375739 [-] on Build GRCm38 104
GenBankmRNA BB640383.1 - 1 - 2 242 Chr. 7 - 73316097 - 73375763 [-] on Build GRCm38 104

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream predicted gene,
30075
Gm30075 -75.1kb Sense Chr. 7 - 73201863 - 73238687 on NCBI Build GRCm38
Downstream chromodomain helicase DNA binding protein 2 Chd2 2810013C04Rik,
2810040A01Rik,
5630401D06Rik,
AI851092,
BC029703,
CHD-2
50.9kb Sense Chr. 7 - 73426652 - 73541746 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700003492.mm.1

Set Membership:

Amplicon length greater than or equal to 101 3' Most Mus musculus 1700 Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700029620.mm.1 NR_040324.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700029620.mm.1 NR_040324.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700029620.mm.1 NR_040325.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700029620.mm.1 NR_040325.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700029620.mm.1 TC0700003492.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700029619.mm.1 NR_040324.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700029619.mm.1 NR_040324.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700029619.mm.1 NR_040325.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700029619.mm.1 NR_040325.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700029619.mm.1 TC0700003492.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700029618.mm.1 NR_040324.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700029618.mm.1 NR_040324.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700029618.mm.1 NR_040325.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700029618.mm.1 NR_040325.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700029618.mm.1 TC0700003492.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 751945 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0