This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04260099_g1
See other PKM GE Assays ›
Gene Symbol:
Pkm
Gene Name:
pyruvate kinase, muscle
Gene Aliases:
AA414905, AL024370, AL024424, Pk-2, Pk-3, Pk3, Pkm2
Chromosome Location:
Chr.9: 59656368 - 59679375 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04260099_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pkm
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001253883.1 NP_001240812.1 1 - 2 281 64
NM_011099.3 NP_035229.2 1 - 2 281 64
GenBank mRNA AK083076.1 - 1 - 2 74 64
AK135397.1 - 1 - 2 74 64
AK151724.1 - 1 - 2 147 64
AK153483.1 - 1 - 2 147 64
AK154695.1 - 1 - 2 74 64
AK155110.1 - 1 - 2 74 64
AK155655.1 - 1 - 2 77 64
AK168943.1 - 1 - 2 74 64
AK169872.1 - 1 - 2 53 64
AK171023.1 - 1 - 2 74 64
AK171033.1 - 1 - 2 77 64
AK171106.1 - 1 - 2 78 64
AK200951.1 - 1 - 2 55 64
AK210659.1 - 1 - 2 38 64
BC016619.1 - 1 - 2 41 3598842 64
CB846337.1 - 1 - 2 290 64

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK135397.1,AK151724.1,AK153483.1,AK155110.1,AK155655.1,AK168943.1,AK171023.1,AK171033.1,BC016619.1,NM_001253883.1,NM_011099.3 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pkm

Panther Classification:

Molecular Function -

kinase

Gene Ontology Categories:

Function(s) Process(es)

liver development
glucose metabolic process
glycolytic process
ATP biosynthetic process
metabolic process
programmed cell death
phosphorylation
organ regeneration
pyruvate biosynthetic process
skeletal muscle tissue regeneration
nucleotide binding
magnesium ion binding
catalytic activity
pyruvate kinase activity
protein binding
ATP binding
kinase activity
transferase activity
MHC class II protein complex binding
potassium ion binding
ADP binding
poly(A) RNA binding
metal ion binding

MicroArray Probes:

ABI

Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 354885 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0