This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04242333_sH
See other MIR181A-2 GE Assays ›
Gene Symbol:
Mir181a-2
Gene Name:
microRNA 181a-2
Gene Aliases:
Mirn181, Mirn181a, Mirn181a-2, miR-181, mir-181a, mir-181a-2
Chromosome Location:
Chr.2: 38852735 - 38852810 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04242333_sH
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mir181a-2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_029568.1 1 37 67

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream nuclear receptor subfamily 5,
group A,
member 1
Nr5a1 Ad4BP,
ELP,
ELP-3,
Ftz-F1,
Ftzf1,
SF-1,
SF1
-138.2kb Anti-sense Chr. 2 - 38692656 - 38714560 on NCBI Build GRCm38
Downstream microRNA 181b-2 Mir181b-2 Mirn181b-2,
mir-181b-2
1.0kb Sense Chr. 2 - 38853830 - 38853918 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s)

cellular response to lipopolysaccharide

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.1 ST Array 10471878 - -
Affymetrix Mouse Clariom S Array PSR0200006257.mm.1 NR_029568.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200006257.mm.1 NR_029568.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200006257.mm.1 TC0200000711.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0