This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04241090_s1
See other MIR3098 GE Assays ›
Gene Symbol:
Mir3098
Gene Name:
microRNA 3098
Gene Aliases:
mmu-mir-3098
Chromosome Location:
Chr.6: 125195792 - 125195878 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04241090_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mir3098
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_037281.1 1 44 77
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AV381250.2 - 1 199 Chr. 6 - 125195647 - 125196263 [+] on Build GRCm38 77
GenBankmRNA BE626205.1 - 1 Chr. 6 - 125195608 - 125196105 [-] on Build GRCm38 77

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream mitochondrial ribosomal protein L51 Mrpl51 2610511M02Rik,
CDA09,
HSPC241,
Mrp64
-1.4kb Sense Chr. 6 - 125192200 - 125194392 on NCBI Build GRCm38
Downstream vesicle-associated membrane protein 1 Vamp1 Syb-1,
Syb1,
VAMP-1,
lew
19.7kb Sense Chr. 6 - 125215581 - 125222306 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200039928.mm.1 NR_037281.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200039928.mm.1 TC0200004648.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0