This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04241068_s1
See other MIR3067 GE Assays ›
Gene Symbol:
Mir3067
Gene Name:
microRNA 3067
Gene Aliases:
mir-3067, mmu-mir-3067
Chromosome Location:
Chr.12: 81166151 - 81166234 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04241068_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mir3067
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_037227.1 1 46 78

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream SPARC related modular calcium binding 1 Smoc1 2600002F22Rik,
AI848508,
SRG
Sense Chr. 12 - 81026808 - 81186414 on NCBI Build GRCm38
Downstream solute carrier family 8 (sodium/calcium exchanger),
member 3
Slc8a3 AW742262,
Ncx3
31.7kb Anti-sense Chr. 12 - 81197915 - 81333728 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1200005339.mm.1 NR_037227.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200005339.mm.1 NR_037227.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200005339.mm.1 TC1200000752.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0