This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04240901_s1
See other MIR1962 GE Assays ›
Gene Symbol:
Mir1962
Gene Name:
microRNA 1962
Gene Aliases:
mir-1962, mmu-mir-1962
Chromosome Location:
Chr.7: 135566162 - 135566282 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04240901_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mir1962
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_035488.1 1 60 63

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream protein tyrosine phosphatase,
receptor type,
E
Ptpre PTPe,
PTPepsilon,
RPTPepsilon
Sense Chr. 7 - 135537645 - 135686294 on NCBI Build GRCm38
Downstream RIKEN cDNA 5830432E09 gene 5830432E09Rik 82.2kb Anti-sense Chr. 7 - 135648457 - 135652314 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700017224.mm.1 NR_035488.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700017224.mm.1 NR_035488.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700017224.mm.1 TC0700001961.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0