This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04240879_s1
See other MIR1931 GE Assays ›
Gene Symbol:
Mir1931
Gene Name:
microRNA 1931
Gene Aliases:
mir-1931, mmu-mir-1931
Chromosome Location:
Chr.10: 93162785 - 93162903 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04240879_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mir1931
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_035452.1 1 62 93
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BQ553652.1 - 1 Chr. 10 - 93162443 - 93163064 [-] on Build GRCm38 93
GenBankmRNA AW557530.1 - 1 Chr. 10 - 93162366 - 93163064 [-] on Build GRCm38 93
GenBankmRNA AU041356.1 - 1 Chr. 10 - 93162484 - 93163064 [-] on Build GRCm38 93

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream cyclin-dependent kinase 17 Cdk17 6430598J10Rik,
Pctk2
Sense Chr. 10 - 93160876 - 93250688 on NCBI Build GRCm38
Downstream ELK3,
member of ETS oncogene family
Elk3 D430049E23Rik,
Erp,
Etrp,
Net,
Sap-2
84.5kb Anti-sense Chr. 10 - 93247415 - 93311167 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000008514.mm.1 NR_035452.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000008514.mm.1 NR_035452.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000008514.mm.1 TC1000001125.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0