This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04240872_s1
See other MIR1904 GE Assays ›
Gene Symbol:
Mir1904
Gene Name:
microRNA 1904
Gene Aliases:
Mirn1904, mmu-mir-1904
Chromosome Location:
Chr.13: 109903809 - 109903888 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04240872_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mir1904
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_035442.1 1 39 73

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream microRNA 582 Mir582 Mirn582,
mir-582,
mmu-mir-582
-579.0kb Sense Chr. 13 - 109324744 - 109324824 on NCBI Build GRCm38
Downstream RAB3C,
member RAS oncogene family
Rab3c 2700062I01Rik,
3110015B08Rik,
3110037E15Rik,
AI850886
150.3kb Anti-sense Chr. 13 - 110054187 - 110281041 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1300007762.mm.1 NR_035442.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300007762.mm.1 NR_035442.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300007762.mm.1 TC1300001209.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0