This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04240861_s1
See other MIR1192 GE Assays ›
Gene Symbol:
Mir1192
Gene Name:
microRNA 1192
Gene Aliases:
Mirn1192, mmu-mir-1192
Chromosome Location:
Chr.19: 23149431 - 23149551 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04240861_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mir1192
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_035423.1 1 55 99

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream Kruppel-like factor 9 Klf9 2310051E17Rik,
AA589643,
BTEB-1,
Bteb1,
ENSMUSG00000055440,
Gm9971
Sense Chr. 19 - 23141226 - 23166911 on NCBI Build GRCm38
Downstream structural maintenance of chromosomes 5 Smc5 MLZ-453,
SMC-5,
Smc5l1,
mKIAA0594,
mSMC5
56.9kb Anti-sense Chr. 19 - 23206441 - 23273908 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1900003568.mm.1 NR_035423.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900003568.mm.1 NR_035423.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900003568.mm.1 TC1900000374.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0