This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04239381_s1
See other B930059L03RIK GE Assays ›
Gene Symbol:
B930059L03Rik
Gene Name:
RIKEN cDNA B930059L03 gene
Gene Aliases:
-
Chromosome Location:
Chr.12: 110591374 - 110592679 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04239381_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: B930059L03Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_033340.1 1 347 90
GenBank mRNA AK047421.1 - 1 348 90
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BX516257.1 - 1 338 Chr. 12 - 110591384 - 110591903 [+] on Build GRCm38 90

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream protein phosphatase 2,
regulatory subunit B',
gamma
Ppp2r5c 2610043M05Rik,
2700063L20Rik,
AI060890,
AW545884,
C85228,
D12Bwg0916e,
mKIAA0044
-8.3kb Sense Chr. 12 - 110447179 - 110583075 on NCBI Build GRCm38
Downstream dynein cytoplasmic 1 heavy chain 1 Dync1h1 9930018I23Rik,
AI894280,
DHC1,
DHC1a,
DNCL,
Dnchc1,
Dnec1,
Dnecl,
Loa,
MAP1C,
P22,
Swl,
mKIAA0325
8.7kb Sense Chr. 12 - 110601395 - 110666944 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1444000_PM_at - -
Affymetrix Mouse Clariom S Array PSR1200008469.mm.1 NR_033340.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200008469.mm.1 NR_033340.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200008469.mm.1 TC1200001209.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200008470.mm.1 NR_033340.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200008470.mm.1 NR_033340.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200008470.mm.1 TC1200001209.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200008471.mm.1 NR_033340.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200008471.mm.1 NR_033340.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200008471.mm.1 TC1200001209.mm.1 MTA TC
Murine Genome U74v2 Set 138092_at - -
Murine Genome U74v2 Set 138092_PM_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 361618 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 103450148 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0