This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04239159_s1
See other D330050I16RIK GE Assays ›
Gene Symbol:
D330050I16Rik
Gene Name:
RIKEN cDNA D330050I16 gene
Gene Aliases:
-
Chromosome Location:
Chr.19: 5388336 - 5390069 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04239159_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: D330050I16Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_033224.1 1 1354 72

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream squamous cell carcinoma antigen recognized by T cells 1 Sart1 U5-110K Anti-sense Chr. 19 - 5377523 - 5389420 on NCBI Build GRCm38
Downstream RIKEN cDNA 4930481A15 gene 4930481A15Rik 16.7kb Sense Chr. 19 - 5406815 - 5422847 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1900000784.mm.1 NR_033224.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900000784.mm.1 NR_033224.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900000784.mm.1 TC1900000063.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0