This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04238288_s1
See other MIR483 GE Assays ›
Gene Symbol:
Mir483
Gene Name:
microRNA 483
Gene Aliases:
Mirn483, mir-483, mmu-mir-483
Chromosome Location:
Chr.7: 142654924 - 142654996 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04238288_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mir483
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_030251.1 1 33 69

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream insulin-like growth factor 2 Igf2 AL033362,
Igf-2,
Igf-II,
M6pr,
Mpr,
Peg2
Sense Chr. 7 - 142650768 - 142662270 on NCBI Build GRCm38
Downstream insulin-like growth factor 2,
opposite strand
Igf2os Igf2as,
Peg8
4.7kb Anti-sense Chr. 7 - 142659693 - 142670356 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s)

response to oxygen levels

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.1 ST Array 10569356 - -
Affymetrix Mouse Clariom S Array PSR0700038567.mm.1 NR_030251.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700038567.mm.1 NR_030251.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700038567.mm.1 TC0700004572.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0