This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04238264_s1
See other MIR133B GE Assays ›
Gene Symbol:
Mir133b Linc-md1
Gene Name:
microRNA 133b
long intergenic non-coding RNA muscle differentiation 1
Gene Aliases:
Mirn133b, mir-133b
-
Chromosome Location:
Chr.1: 20682769 - 20682887 on Build GRCm38
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
Assay ID Mm04238264_s1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mir133b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_029902.1 1 51 73
Gene Symbol: Linc-md1
Interrogated Sequence Translated Protein RNADB ID Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK132542.1 - LIT3444
4831414J02Rik
1 3800 73
JN699052.1 -
3 382 73

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Inventoried Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s)

positive regulation of gene expression
negative regulation of neuron maturation
response to oxygen levels

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.1 ST Array 10345030 - -
Affymetrix Mouse Clariom S Array PSR0100000890.mm.1 NR_029902.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100000890.mm.1 NR_029902.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100000890.mm.1 TC0100000125.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-655dcf6cc8-78hlp:80/100.66.128.142:80.
git-commit: 0d764ad91c70359c551b4d51a80efc6847a0ee54
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0