This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04238261_s1
See other MIR383 GE Assays ›
Gene Symbol:
Mir383
Gene Name:
microRNA 383
Gene Aliases:
Mirn383, mir-383, mmu-mir-383
Chromosome Location:
Chr.8: 38252133 - 38252202 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04238261_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mir383
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_029884.1 1 34 58

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream sarcoglycan zeta Sgcz C230085N17Rik Sense Chr. 8 - 37517840 - 38661508 on NCBI Build GRCm38
Downstream tumor suppressor candidate 3 Tusc3 AU022242,
BC003311,
N33
753.7kb Anti-sense Chr. 8 - 39157260 - 39165114 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s)

cellular response to lipopolysaccharide

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.1 ST Array 10578262 - -
Affymetrix Mouse Clariom S Array PSR0800016114.mm.1 NR_029884.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800016114.mm.1 NR_029884.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800016114.mm.1 TC0800002091.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0