This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04238227_s1
See other MIR210 GE Assays ›
Gene Symbol:
Mir210
Gene Name:
microRNA 210
Gene Aliases:
Mirn210, mir-210, mmu-mir-210
Chromosome Location:
Chr.7: 141221384 - 141221493 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04238227_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mir210
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_029793.1 1 54 81

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream Ras association (RalGDS/AF-6) domain family (N-terminal) member 7 Rassf7 2400009B11Rik,
AW210608
-2.7kb Anti-sense Chr. 7 - 141215560 - 141218658 on NCBI Build GRCm38
Downstream PHD and ring finger domains 1 Phrf1 AA673488 6.8kb Anti-sense Chr. 7 - 141228263 - 141262751 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s)

BMP signaling pathway
negative regulation of activin receptor signaling pathway
positive regulation of osteoblast differentiation
cellular response to lipopolysaccharide

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.1 ST Array 10569100 - -
Affymetrix Mouse Clariom S Array PSR0700038010.mm.1 NR_029793.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700038010.mm.1 NR_029793.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700038010.mm.1 TC0700004528.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0