This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04238204_s1
See other MIR326 GE Assays ›
Gene Symbol:
Mir326
Gene Name:
microRNA 326
Gene Aliases:
Mirn326, mir-326, mmu-mir-326
Chromosome Location:
Chr.7: 99552269 - 99552363 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04238204_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mir326
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_029760.1 1 43 48

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream ribosomal protein S3 Rps3 D7Ertd795e,
Rs_3
-68.6kb Anti-sense Chr. 7 - 99477896 - 99483709 on NCBI Build GRCm38
Downstream predicted gene 4980 Gm4980 6430704N06,
EG245190,
Tpbgl
71.7kb Anti-sense Chr. 7 - 99624067 - 99627103 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s)

regulation of gene expression
positive regulation of T-helper 17 cell differentiation

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.1 ST Array 10555254 - -
Affymetrix Mouse Clariom S Array PSR0700011806.mm.1 NR_029760.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700011806.mm.1 NR_029760.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700011806.mm.1 TC0700001392.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0