This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04238190_s1
See other MIR26B GE Assays ›
Gene Symbol:
Mir26b
Gene Name:
microRNA 26b
Gene Aliases:
Mirn26b, mir-26b, mmu-mir-26b
Chromosome Location:
Chr.1: 74394310 - 74394394 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04238190_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mir26b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_029743.1 1 43 68

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream CTD (carboxy-terminal domain,
RNA polymerase II,
polypeptide A) small phosphatase 1
Ctdsp1 GIP,
NLIIF,
Nif3,
SCP1
Sense Chr. 1 - 74391609 - 74397285 on NCBI Build GRCm38
Downstream villin 1 Vil1 Vil 14.8kb Sense Chr. 1 - 74409191 - 74435560 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s)

negative regulation of protein phosphorylation
gene silencing by miRNA
response to oxygen levels
cellular response to lipopolysaccharide
cellular response to growth factor stimulus
negative regulation of G1/S transition of mitotic cell cycle

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.1 ST Array 10347362 - -
Affymetrix Mouse Clariom S Array PSR0100005126.mm.1 NR_029743.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100005126.mm.1 NR_029743.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100005126.mm.1 TC0100000634.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0