This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04238165_s1
See other MIR130B GE Assays ›
Gene Symbol:
Mir130b
Gene Name:
microRNA 130b
Gene Aliases:
Mirn130b, mir-130b
Chromosome Location:
Chr.16: 17124061 - 17124142 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04238165_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mir130b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_029659.1 1 40 61

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream peptidylprolyl isomerase (cyclophilin)-like 2 Ppil2 0610009L05Rik,
1700016N17Rik,
4921520K19Rik,
4930511F14Rik,
AA589416,
C130078A06Rik,
CYC4
-12.8kb Sense Chr. 16 - 17086555 - 17111263 on NCBI Build GRCm38
Downstream microRNA 301b Mir301b Mirn301b,
mir-301b
258.0 Sense Chr. 16 - 17124400 - 17124496 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.1 ST Array 10438094 - -
Affymetrix Mouse Clariom S Array PSR1600010358.mm.1 NR_029659.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600010358.mm.1 NR_029659.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600010358.mm.1 TC1600001278.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-655dcf6cc8-78hlp:80/100.66.128.142:80.
git-commit: 0d764ad91c70359c551b4d51a80efc6847a0ee54
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0