This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04238141_s1
See other MIR202 GE Assays ›
Gene Symbol:
Mir202 Gm2044
Gene Name:
microRNA 202
predicted gene 2044
Gene Aliases:
Mirn202, mir-202, mmu-mir-202
-
Chromosome Location:
Chr.7: 139957689 - 139957760 on Build GRCm38
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
Assay ID Mm04238141_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mir202
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_029589.1 1 37 72
Gene Symbol: Gm2044
Interrogated Sequence Translated Protein RNADB ID Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK144366.1 - LIT3443
G630041I22Rik
193 72
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BB873022.1 - 2 193 Chr. 7 - 139957561 - 139958039 [-] on Build GRCm38 72
GenBankmRNA CB572889.1 - 2 195 Chr. 7 - 139957207 - 139958041 [-] on Build GRCm38 72

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream uncharacterized LOC102635948 LOC102635948 Anti-sense Chr. 7 - 139950001 - 139978980 on NCBI Build GRCm38
Downstream RIKEN cDNA 6430531B16 gene 6430531B16Rik Gm1093 14.5kb Sense Chr. 7 - 139972303 - 139978755 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s)

cellular response to lipopolysaccharide

MicroArray Probes:

Agilent Affymetrix

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2083368 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2083368 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.1 ST Array 10568863 - -
Mouse Genome 430 2.0 Array 1447352_PM_at - -
Mouse Genome 430 2.0 Array 1447352_at - -
Affymetrix Mouse Clariom S Array PSR0700037619.mm.1 NR_029589.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700037619.mm.1 NR_029589.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700037619.mm.1 TC0700004485.mm.1 MTA TC
Mouse Expression Set 430 1447352_at - -
Mouse Expression Set 430 1447352_PM_at - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0