This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04238126_s1
See other MIR151 GE Assays ›
Gene Symbol:
Mir151
Gene Name:
microRNA 151
Gene Aliases:
Mirn151, mir-151, mmu-mir-151
Chromosome Location:
Chr.15: 73254815 - 73254882 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04238126_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mir151
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_029561.1 1 34 61

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream argonaute RISC catalytic subunit 2 Ago2 1110029L17Rik,
2310051F07Rik,
AI225898,
AL022874,
AW546247,
ENSMUSG00000072493,
Eif2c2,
Gerp95,
Gm10365,
mKIAA4215
-69.9kb Sense Chr. 15 - 73101625 - 73184947 on NCBI Build GRCm38
Downstream DENN/MADD domain containing 3 Dennd3 AI447457,
E030003N15Rik
257.7kb Anti-sense Chr. 15 - 73512560 - 73572242 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s)

cellular response to amino acid stimulus

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.1 ST Array 10429385 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0