This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04238125_s1
See other MIR150 GE Assays ›
Gene Symbol:
Mir150
Gene Name:
microRNA 150
Gene Aliases:
Mirn150, mir-150, mmu-mir-150
Chromosome Location:
Chr.7: 45121757 - 45121821 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04238125_s1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mir150
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_029560.1 1 34 61

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s)

immunoglobulin production
regulation of gene expression
regulation of B cell differentiation
homeostasis of number of cells
cellular response to lipopolysaccharide

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.1 ST Array 10552843 - -
Affymetrix Mouse Clariom S Array PSR0700007084.mm.1 NR_029560.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700007084.mm.1 NR_029560.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700007084.mm.1 TC0700000829.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0