This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04237573_m1
See other D130020L05RIK GE Assays ›
Gene Symbol:
D130020L05Rik
Gene Name:
RIKEN cDNA D130020L05 gene
Gene Aliases:
9430007L23Rik
Chromosome Location:
Chr.12: 101082451 - 101088927 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04237573_m1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: D130020L05Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_038049.1 1 - 2 1573 96
GenBank mRNA BX512124.1 - 1 - 2 112 96
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA CF198210.1 - 1 - 2 80 Chr. 12 - 101083945 - 101088175 [+] on Build GRCm38 96
GenBankmRNA BG100603.1 - 1 - 2 196 Chr. 12 - 101083828 - 101088183 [+] on Build GRCm38 96
GenBankmRNA CA458682.1 - 101 Chr. - - [-] on Build 96
GenBankmRNA AA289599.1 - 1 - 2 272 Chr. 12 - 101083749 - 101088158 [+] on Build GRCm38 96
GenBankmRNA AA404206.1 - 1 - 2 111 Chr. 12 - 101083913 - 101088178 [+] on Build GRCm38 96

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream SMEK homolog 1,
suppressor of mek1 (Dictyostelium)
Smek1 1110034C04Rik,
BC064465,
mKIAA2010
Anti-sense Chr. 12 - 101039409 - 101083702 on NCBI Build GRCm38
Downstream kinesin family member 4,
pseudogene
Kif4-ps 4930463M05Rik,
Kif4b
56.7kb Sense Chr. 12 - 101145609 - 101149273 on NCBI Build GRCm38

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Mus musculus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0