This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04237571_m1
See other D130020L05RIK GE Assays ›
Gene Symbol:
D130020L05Rik
Gene Name:
RIKEN cDNA D130020L05 gene
Gene Aliases:
9430007L23Rik
Chromosome Location:
Chr.12: 101082451 - 101088927 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04237571_m1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: D130020L05Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_038047.1 2 - 3 1786 95
GenBank mRNA BU939156.1 - 2 - 3 498 95
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AA183837.1 - 2 - 3 265 Chr. 12 - 101083970 - 101086294 [+] on Build GRCm38 95
GenBankmRNA CB271103.1 - 1 - 2 222 Chr. 12 - 101084281 - 101088172 [+] on Build GRCm38 95

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream SMEK homolog 1,
suppressor of mek1 (Dictyostelium)
Smek1 1110034C04Rik,
BC064465,
mKIAA2010
-582.0 Anti-sense Chr. 12 - 101039409 - 101083702 on NCBI Build GRCm38
Downstream kinesin family member 4,
pseudogene
Kif4-ps 4930463M05Rik,
Kif4b
59.3kb Sense Chr. 12 - 101145609 - 101149273 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1200001000.mm.1

Set Membership:

Amplicon length between 86 and 100 3' Most Mus musculus Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1200007021.mm.1 NR_038047.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200007021.mm.1 NR_038047.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200007021.mm.1 TC1200001000.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0