This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04237550_m1
See other E130307A14RIK GE Assays ›
Gene Symbol:
E130307A14Rik
Gene Name:
RIKEN cDNA E130307A14 gene
Gene Aliases:
-
Chromosome Location:
Chr.10: 39621411 - 39732007 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04237550_m1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: E130307A14Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_038037.1 2 - 3 346 151
GenBank mRNA AK164126.1 - 2 - 3 306 151
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY729658.1 - 2 - 3 306 Chr. 10 - 39622406 - 39731967 [-] on Build GRCm38 151
GenBankmRNA BQ877802.1 - 294 Chr. - - [-] on Build 151
GenBankmRNA AU035830.1 - 2 - 3 326 Chr. 10 - 39622363 - 39731987 [-] on Build GRCm38 151
GenBankmRNA BE649737.1 - 2 - 3 177 Chr. 10 - 39622382 - 39731838 [-] on Build GRCm38 151
GenBankmRNA BY266180.1 - 2 - 3 205 Chr. 10 - 39622514 - 39731865 [-] on Build GRCm38 151

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream predicted gene 16364 Gm16364 OTTMUSG00000034131 -7.5kb Sense Chr. 10 - 39552967 - 39613898 on NCBI Build GRCm38
Downstream REV3-like,
catalytic subunit of DNA polymerase zeta RAD54 like (S. cerevisiae)
Rev3l Rev,
Rev3,
Sez4
50.6kb Anti-sense Chr. 10 - 39732099 - 39875210 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000002025.mm.1

Set Membership:

Amplicon length greater than or equal to 101 3' Most Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 849316 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 249905 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 600642 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 620249 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 529904 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 955918 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 740589 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 853843 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 37418 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 40250 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 733754 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 802069 AK031067.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 802069 BC094578.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 836786 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 626120 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 92683 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 356251 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 805639 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 633597 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 784357 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 691259 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 851568 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 831919 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 822672 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 646385 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 127421 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 946664 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 530191 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 685657 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 877826 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 41344 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 34243 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 505254 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 188188 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 793574 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 86417 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 670883 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 235038 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 116163 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 166007 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 62345 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 134014 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 143650 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 82253 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 17231 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 42281 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P132584 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P136325 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P131103 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P352099 AK049799.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P352099 AK049799.1 mouse_transcriptome
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P136325 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07034360 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P131103 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P132584 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07034377 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07034445 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07034396 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02107291 - -
Whole Mouse Genome G4122A A_52_P352099 AK049799.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P352099 AK049799.1 mouse_transcriptome
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P136325 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P131103 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P132584 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P02107291 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08348428 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080116 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080225 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080126 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080226 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080266 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08523186 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080079 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080217 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080203 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080177 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080086 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080084 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080172 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080182 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080088 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080179 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08216570 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080153 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080273 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080063 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080069 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080095 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080065 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080104 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080169 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080212 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080165 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080288 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080150 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080155 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080262 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080173 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08857337 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08597367 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080174 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08417933 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080151 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080089 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08523187 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080285 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080073 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080208 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080180 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080209 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080269 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080197 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080068 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08229885 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080121 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080224 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08229886 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080281 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080206 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080194 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080082 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080282 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080083 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080125 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080286 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080123 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08335585 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080215 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080211 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080193 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080129 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080287 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080157 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080227 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08735886 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080161 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080087 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08597371 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08317193 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080202 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080210 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080220 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080271 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080201 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08523181 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080092 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080096 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080118 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080163 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080277 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080124 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080072 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080102 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080260 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08590600 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080075 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080222 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080106 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080090 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27080176 - -
Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1434723_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1443261_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1440174_PM_at - -
Mouse Genome 430 2.0 Array 1440174_at - -
Mouse Genome 430 2.0 Array 1434723_PM_at - -
Mouse Genome 430 2.0 Array 1443261_at - -
Mouse Genome 430 2.0 Array 1434723_at - -
Mouse Genome 430 2.0 Array 1443261_PM_at - -
Mouse Genome 430 2.0 Array 1440174_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 327744_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 327744_at - -
Affymetrix Mouse Clariom S Array PSR1000014354.mm.1 NR_038037.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000014354.mm.1 NR_038037.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000014354.mm.1 TC1000002025.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000014356.mm.1 NR_038037.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000014356.mm.1 NR_038037.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000014356.mm.1 TC1000002025.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000014396.mm.1 NR_038037.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000014396.mm.1 NR_038037.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000014396.mm.1 TC1000002025.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000014351.mm.1 NR_038037.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000014351.mm.1 NR_038037.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000014351.mm.1 TC1000002025.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000014355.mm.1 NR_038037.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000014355.mm.1 NR_038037.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000014355.mm.1 TC1000002025.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000014406.mm.1 NR_038037.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000014406.mm.1 NR_038037.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000014406.mm.1 TC1000002025.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000014353.mm.1 NR_038037.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000014353.mm.1 NR_038037.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000014353.mm.1 TC1000002025.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000014403.mm.1 NR_038037.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000014403.mm.1 NR_038037.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000014403.mm.1 TC1000002025.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000014402.mm.1 NR_038037.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000014402.mm.1 NR_038037.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000014402.mm.1 TC1000002025.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000014405.mm.1 NR_038037.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000014405.mm.1 NR_038037.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000014405.mm.1 TC1000002025.mm.1 MTA TC
Mouse Expression Set 430 1443261_PM_at - -
Mouse Expression Set 430 1443261_at - -
Mouse Expression Set 430 1440174_at - -
Mouse Expression Set 430 1434723_at - -
Mouse Expression Set 430 1434723_PM_at - -
Mouse Expression Set 430 1440174_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1434723_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1434723_at - -
Affymetrix HT Mouse Genome 430B Array 1443261_at - -
Affymetrix HT Mouse Genome 430B Array 1440174_at - -
Affymetrix HT Mouse Genome 430B Array 1440174_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1443261_PM_at - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 2370204 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0