This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04236247_m1
See other AIRN GE Assays ›
Gene Symbol:
Airn
Gene Name:
antisense Igf2r RNA
Gene Aliases:
2810051F02Rik, 2810434M15Rik, AI256653, AI597500, AW049873, Air, B930018I07Rik, D17Ertd663e, IGF2RAS
Chromosome Location:
Chr.17: 12741311 - 12859884 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04236247_m1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Airn
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_027772.1 1 - 2 112 63
GenBank mRNA BF729958.1 - 1 - 2 93 63
DQ275619.1 - 1 - 2 44 63
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY102556.1 - 1 - 2 51 Chr. 17 - 12741374 - 12830122 [+] on Build GRCm38 63

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream solute carrier family 22 (organic cation transporter),
member 1
Slc22a1 Lx1,
Oct1,
Orct,
Orct1
-65.5kb Anti-sense Chr. 17 - 12648874 - 12675838 on NCBI Build GRCm38
Downstream MAS1 oncogene Mas1 Mas-1,
Mgra
8.7kb Anti-sense Chr. 17 - 12838841 - 12868145 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1700000127.mm.1

Set Membership:

Amplicon length less than or equal to 70 3' Most Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

regulation of gene expression by genetic imprinting
regulation of histone H3-K9 methylation
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700001043.mm.1 NR_027772.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700001043.mm.1 NR_027772.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700001043.mm.1 TC1700000127.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0