This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04212699_m1
See other TMEM241 GE Assays ›
Gene Symbol:
Tmem241
Gene Name:
transmembrane protein 241
Gene Aliases:
6030446N20Rik
Chromosome Location:
Chr.18: 11980136 - 12121543 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04212699_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tmem241
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001289666.1 NP_001276595.1 2 - 3 201 75
NM_001289667.1 NP_001276596.1 2 - 3 201 75
NM_178801.5 NP_848916.2 2 - 3 201 75
NR_110356.1 2 - 3 201 75
GenBank mRNA AK031517.1 - 2 - 3 195 75
AK038419.1 - 2 - 3 179 75
AK038472.1 - 2 - 3 146 75
AK051727.1 - 2 - 3 202 75
AK144963.1 - 2 - 3 203 75
BC094382.1 - 2 - 3 117 30605060 75
BC137621.1 - 2 - 3 131 8860640 75
BC144788.1 - 2 - 3 132 75

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK038472.1, NM_001289667.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tmem241

Panther Classification:

Molecular Function -

secondary carrier transporter

Gene Ontology Categories:

Function(s) Process(es)

protein glycosylation
biological_process
anion transmembrane transport
GDP-mannose transmembrane transport
molecular_function
nucleotide-sugar transmembrane transporter activity
antiporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1800007362.mm.1 NM_001289666.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800007362.mm.1 NM_001289667.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800007362.mm.1 NM_178801.5 Refseq
Affymetrix Mouse Clariom S Array PSR1800007362.mm.1 NR_110356.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800007362.mm.1 TC1800000990.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0