This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04208596_m1
See other TNFAIP8L1 GE Assays ›
Gene Symbol:
Tnfaip8l1
Gene Name:
tumor necrosis factor, alpha-induced protein 8-like 1
Gene Aliases:
2600017J23Rik, TIPE1
Chromosome Location:
Chr.17: 56162491 - 56173955 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04208596_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tnfaip8l1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_025566.3 NP_079842.2 1 - 2 119 77
GenBank mRNA AK090348.1 - 1 - 2 121 77
AK136962.1 - 1 - 2 81 77
AK150678.1 - 1 - 2 110 77
AK153177.1 - 1 - 2 105 77
BY733616.1 - 1 - 2 119 77

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK090348.1, AK136962.1, AK150678.1, AK153177.1, NM_025566.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tnfaip8l1

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of TOR signaling
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700009559.mm.1 NM_025566.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700009559.mm.1 NM_025566.3 Refseq
Affymetrix Mouse Clariom S Array PSR1700009559.mm.1 TC1700000999.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1700009558.mm.1 NM_025566.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700009558.mm.1 NM_025566.3 Refseq
Affymetrix Mouse Clariom S Array PSR1700009558.mm.1 TC1700000999.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1700009557.mm.1 NM_025566.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700009557.mm.1 NM_025566.3 Refseq
Affymetrix Mouse Clariom S Array PSR1700009557.mm.1 TC1700000999.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0