This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04208479_g1
See other NDUFA4 GE Assays ›
Gene Symbol:
Ndufa4
Gene Name:
NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 4
Gene Aliases:
MLRQ
Chromosome Location:
Chr.6: 11900373 - 11907446 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04208479_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ndufa4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010886.2 NP_035016.1 1 - 2 125 75
GenBank mRNA AK005084.1 - 1 - 2 130 75
AK008357.1 - 1 - 2 122 75
AK008506.1 - 1 - 2 116 75
AK076036.1 - 1 - 2 126 75
AK154084.1 - 1 - 2 109 75
BC011114.1 - 1 - 2 88 3483028 75

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ndufa4

Panther Classification:

Molecular Function -

oxidoreductase

Gene Ontology Categories:

Function(s) Process(es)

transport
oxidation-reduction process
hydrogen ion transmembrane transport
cytochrome-c oxidase activity
protein complex binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600014659.mm.1 NM_010886.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600014659.mm.1 NM_010886.2 Refseq
Affymetrix Mouse Clariom S Array PSR0600014659.mm.1 TC0600001862.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600014661.mm.1 NM_010886.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600014661.mm.1 NM_010886.2 Refseq
Affymetrix Mouse Clariom S Array PSR0600014661.mm.1 TC0600001862.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600014660.mm.1 NM_010886.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600014660.mm.1 NM_010886.2 Refseq
Affymetrix Mouse Clariom S Array PSR0600014660.mm.1 TC0600001862.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0