This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04208278_s1
See other IFNA4 GE Assays ›
Gene Symbol:
Ifna4
Gene Name:
interferon alpha 4
Gene Aliases:
Ifa4
Chromosome Location:
Chr.4: 88841816 - 88842459 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04208278_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ifna4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010504.2 NP_034634.1 1 48 102
GenBank mRNA BC119349.1 - 1 48 8734098 102
BC119351.1 - 1 48 8734100 102

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC119349.1, BC119351.1, NM_010504.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ifna4

Panther Classification:

Molecular Function -

interferon superfamily

Gene Ontology Categories:

Function(s) Process(es)

adaptive immune response
T cell activation involved in immune response
natural killer cell activation involved in immune response
defense response
humoral immune response
cytokine-mediated signaling pathway
B cell differentiation
positive regulation of peptidyl-serine phosphorylation of STAT protein
B cell proliferation
response to exogenous dsRNA
defense response to virus
cytokine activity
cytokine receptor binding
type I interferon receptor binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400006106.mm.1 NM_010504.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400006106.mm.1 NM_010504.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400006106.mm.1 TC0400000908.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400006107.mm.1 NM_010504.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400006107.mm.1 NM_010504.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400006107.mm.1 TC0400000908.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0