This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04208212_g1
See other PRDX2 GE Assays ›
Gene Symbol:
Prdx2
Gene Name:
peroxiredoxin 2
Gene Aliases:
AL022839, Band-8, NkefB, PRP, PrxII, TDX1, TPx, TPx-B, TR, TSA, Tdpx1, Torin
Chromosome Location:
Chr.8: 84969648 - 84974317 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04208212_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Prdx2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011563.5 NP_035693.3 2 - 3 220 102
GenBank mRNA AK005225.1 - 2 - 3 248 102
AK008433.1 - 2 - 3 225 102
AK014765.1 - 1 - 2 178 102
AK088280.1 - 2 - 3 239 102
BC002034.1 - 2 - 3 228 3485635 102
BC081454.1 - 2 - 3 139 6818396 102
BC086783.1 - 1 - 2 106 5700069 102
U20611.1 - 2 - 3 182 102
U51679.1 - 2 - 3 157 102
X82067.1 - 2 - 3 189 102

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Prdx2

Panther Classification:

Molecular Function -

peroxidase

Gene Ontology Categories:

Function(s) Process(es)

activation of MAPK activity
respiratory burst involved in inflammatory response
response to oxidative stress
regulation of hydrogen peroxide metabolic process
removal of superoxide radicals
positive regulation of blood coagulation
negative regulation of lipopolysaccharide-mediated signaling pathway
negative regulation of NF-kappaB transcription factor activity
response to lipopolysaccharide
cellular response to oxidative stress
T cell proliferation
hydrogen peroxide metabolic process
hydrogen peroxide catabolic process
regulation of apoptotic process
negative regulation of neuron apoptotic process
negative regulation of T cell differentiation
thymus development
homeostasis of number of cells
oxidation-reduction process
negative regulation of reactive oxygen species metabolic process
negative regulation of extrinsic apoptotic signaling pathway
negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
peroxidase activity
protein binding
thioredoxin peroxidase activity
selenium binding
antioxidant activity
oxidoreductase activity
peroxiredoxin activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800007403.mm.1 NM_011563.6 Refseq
Affymetrix Mouse Clariom S Array PSR0800007403.mm.1 TC0800000974.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0