This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04208097_gH
See other PIRA4 GE Assays ›
Gene Symbol:
Pira4 Gm14548 Pira1 Pira11 Pira2 Pira5 Pira6 Pira7 Pirb
Gene Name:
paired-Ig-like receptor A4
predicted gene 14548
paired-Ig-like receptor A1
paired-Ig-like receptor A11
paired-Ig-like receptor A2
paired-Ig-like receptor A5
paired-Ig-like receptor A6
paired-Ig-like receptor A7
paired Ig-like receptor B
Gene Aliases:
6M7
OTTMUSG00000017169
6M21, Ly89, PIR-A1, Pir
Gm14548, p91C
6M23
7M4
12M1, Pira10, p91B
16M2
Gp91, LIR-3, Lilrb3, PIR-B
Chromosome Location:
-
Chromosome Location:
Chr.7: 3884201 - 3898159 on Build GRCm38
Chromosome Location:
Chr.7: 3731633 - 3739870 on Build GRCm38
Chromosome Location:
-
Chromosome Location:
Chr.7: 3836810 - 3845099 on Build GRCm38
Chromosome Location:
-
Chromosome Location:
Chr.7: 4274124 - 4284827 on Build GRCm38
Chromosome Location:
-
Chromosome Location:
Chr.7: 3712505 - 3720456 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
-
-
-
-
-
-
-
Assay ID Mm04208097_gH
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pira4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011091.1 NP_035221.1 1557 61
GenBank mRNA U96685.1 - 1557 61
Gene Symbol: Gm14548
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001166672.1 NP_001160144.1 7 - 8 1557 61
Gene Symbol: Pira1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011087.1 NP_035217.1 1558 61
GenBank mRNA U96682.1 - 1558 61
Gene Symbol: Pira11
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011088.1 NP_035218.1 1558 61
GenBank mRNA AF041036.1 - 1549 61
AF055896.1 - 1549 61
BC103528.2 - 1558 40039374 61
BC103529.1 - 1558 40039371 61
BC139803.1 - 1273 40039373 61
BC150155.1 - 1276 40141323 61
BC150712.1 - 1305 9087623 61
Gene Symbol: Pira2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011089.2 NP_035219.2 7 - 8 1577 61
GenBank mRNA AK137475.1 - 7 - 8 1578 61
AK140776.1 - 7 - 8 1583 61
AK172046.1 - 1578 61
U96683.1 - 1544 61
Gene Symbol: Pira5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA U96686.1 - 7 - 8 1261 61
Gene Symbol: Pira6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001289428.1 NP_001276357.1 1614 61
NM_008848.1 NP_032874.1 1549 61
GenBank mRNA AF041035.1 - 1549 61
Gene Symbol: Pira7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011094.1 NP_035224.1 7 - 8 1585 61
GenBank mRNA U96688.1 - 7 - 8 1585 61
Gene Symbol: Pirb
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011095.2 NP_035225.2 7 - 8 1605 61
GenBank mRNA AF148878.1 - 1551 61
AF148879.1 - 1560 61
AF148880.1 - 7 - 8 1556 61
AF288356.1 - 1555 61
AK142375.1 - 7 - 8 1577 61
AK152232.1 - 6 - 7 1808 61
AK153116.1 - 6 - 7 1808 61
AK161081.1 - 1576 61
AK165255.1 - 7 - 8 1605 61
AK165650.1 - 1734 61
BC026937.1 - 1559 4488338 61
U83172.1 - 7 - 8 1549 61
U96689.1 - 7 - 8 1588 61
U96690.1 - 1540 61
U96691.1 - 1552 61
U96692.1 - 7 - 8 1589 61
U96693.1 - 7 - 8 1605 61

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pira4
Click here to find siRNAs for Gene Gm14548
Click here to find siRNAs for Gene Pira1
Click here to find siRNAs for Gene Pira11
Click here to find siRNAs for Gene Pira2
Click here to find siRNAs for Gene Pira5
Click here to find siRNAs for Gene Pira6
Click here to find siRNAs for Gene Pira7
Click here to find siRNAs for Gene Pirb

Panther Classification:

Molecular Function -

immunoglobulin receptor superfamily

Gene Ontology Categories:

Function(s) Process(es)

biological_process
B cell homeostasis
immune system process
cytokine-mediated signaling pathway
B cell mediated immunity
myeloid dendritic cell differentiation
molecular_function

MicroArray Probes:

ABI

Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 694880 - -
ABI Mouse Genome Survey Microarray 746329 - -
ABI Mouse Genome Survey Microarray 916589 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0