This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04207359_m1
See other CSRP2 GE Assays ›
Gene Symbol:
Csrp2
Gene Name:
cysteine and glycine-rich protein 2
Gene Aliases:
AW551867, Crp2, SmLim
Chromosome Location:
Chr.10: 110920176 - 110939514 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04207359_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Csrp2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_007792.4 NP_031818.3 3 - 4 361 84
GenBank mRNA AF037208.1 - 3 - 4 361 84
AK003328.1 - 3 - 4 360 84
AK010626.1 - 3 - 4 362 84
AK012647.1 - 3 - 4 359 84
BC012663.1 - 3 - 4 346 4210909 84
CK031156.1 - 3 - 4 347 84
D88792.1 - 2 - 3 287 84

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000001284.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Csrp2

Panther Classification:

Molecular Function -

actin or actin-binding cytoskeletal protein

Gene Ontology Categories:

Function(s) Process(es)

multicellular organism development
cell differentiation
protein binding
zinc ion binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000009616.mm.1 NM_007792.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000009616.mm.1 NM_007792.4 Refseq
Affymetrix Mouse Clariom S Array PSR1000009616.mm.1 TC1000001284.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0