This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04207352_m1
See other CITED1 GE Assays ›
Gene Symbol:
Cited1
Gene Name:
Cbp/p300-interacting transactivator with Glu/Asp-rich carboxy-terminal domain 1
Gene Aliases:
AI316840, AU019144, Msg1
Chromosome Location:
Chr.X: 102247378 - 102252181 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04207352_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cited1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_007709.4 NP_031735.1 1 - 2 346 101
GenBank mRNA BC052030.1 - 1 - 2 272 5709775 101

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC052030.1, NM_007709.4 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cited1

Panther Classification:

Molecular Function -

transcription cofactor

Gene Ontology Categories:

Function(s) Process(es)

embryonic axis specification
vasculogenesis
branching involved in ureteric bud morphogenesis
kidney development
placenta development
morphogenesis of an epithelium
negative regulation of mesenchymal to epithelial transition involved in metanephros morphogenesis
transcription, DNA-templated
regulation of transcription, DNA-templated
nucleocytoplasmic transport
apoptotic process
transforming growth factor beta receptor signaling pathway
multicellular organism development
positive regulation of gene expression
cell differentiation
melanocyte differentiation
response to lipopolysaccharide
response to insulin
response to cytokine
response to interferon-gamma
pigmentation
negative regulation of neuron apoptotic process
response to estrogen
negative regulation of osteoblast differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
response to cAMP
SMAD protein signal transduction
labyrinthine layer development
spongiotrophoblast layer development
response to interleukin-1
response to interleukin-2
response to interleukin-4
response to interleukin-6
response to interleukin-9
response to interleukin-11
response to parathyroid hormone
response to transforming growth factor beta
chromatin binding
transcription factor activity, sequence-specific DNA binding
transcription coactivator activity
protein binding
protein C-terminus binding
protein homodimerization activity
transcription regulatory region DNA binding
LBD domain binding
co-SMAD binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0X00017283.mm.1 NM_007709.4 Refseq
Affymetrix Mouse Clariom S Array PSR0X00017283.mm.1 TC0X00002739.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0X00017284.mm.1 NM_007709.4 Refseq
Affymetrix Mouse Clariom S Array PSR0X00017284.mm.1 TC0X00002739.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0X00017278.mm.1 NM_001276466.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00017278.mm.1 NM_001276473.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00017278.mm.1 NM_001276474.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00017278.mm.1 NM_007709.4 Refseq
Affymetrix Mouse Clariom S Array PSR0X00017278.mm.1 TC0X00002739.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0