This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04207240_g1
See other HNRNPC GE Assays ›
Gene Symbol:
Hnrnpc
Gene Name:
heterogeneous nuclear ribonucleoprotein C
Gene Aliases:
AL022939, D14Wsu171e, Hnrpc, Hnrpc1, Hnrpc2, hnRNPC1, hnRNPC2, hnrnp-C
Chromosome Location:
Chr.14: 52073380 - 52104028 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04207240_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hnrnpc
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001170981.1 NP_001164452.1 1 - 2 139 98
NM_001170983.1 NP_001164454.1 1 - 2 139 98
NM_001170984.1 NP_001164455.1 1 - 2 139 98
NM_016884.3 NP_058580.1 1 - 2 139 98
GenBank mRNA AK010845.1 - 1 - 2 118 98
AK011336.1 - 1 - 2 111 98
AK012633.2 - 1 - 2 101 98
AK088245.1 - 1 - 2 116 98
AK088678.1 - 1 - 2 111 98
AK089061.1 - 1 - 2 118 99
AK150399.1 - 1 - 2 116 98
AK151446.1 - 1 - 2 116 98
AK153053.1 - 1 - 2 116 98
AK166590.1 - 1 - 2 117 98
AK168802.1 - 1 - 2 113 98
AK169287.1 - 1 - 2 111 98
BC004706.1 - 1 - 2 101 3499283 98
DV652725.1 - 1 - 2 163 98

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the AK011336.1,AK012633.2,AK088245.1,AK088678.1,AK089061.1,AK153053.1,AK166590.1,AK168802.1,BC004706.1,NM_001170981.1,NM_001170983.1,NM_001170984.1,NM_016884.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Hnrnpc

Gene Ontology Categories:

Function(s) Process(es)

mRNA splicing, via spliceosome
osteoblast differentiation
mRNA processing
RNA splicing
ATP-dependent chromatin remodeling
3'-UTR-mediated mRNA stabilization
nucleotide binding
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II distal enhancer sequence-specific DNA binding
nucleic acid binding
RNA binding
mRNA binding
mRNA 3'-UTR binding
poly(U) RNA binding
nucleosomal DNA binding
identical protein binding
poly(A) RNA binding
N6-methyladenosine-containing RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400015167.mm.1 NM_001170981.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400015167.mm.1 NM_001170981.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400015167.mm.1 NM_001170983.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400015167.mm.1 NM_001170983.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400015167.mm.1 NM_001170984.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400015167.mm.1 NM_001170984.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400015167.mm.1 NM_016884.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400015167.mm.1 NM_016884.3 Refseq
Affymetrix Mouse Clariom S Array PSR1400015167.mm.1 TC1400002023.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400015174.mm.1 NM_001170981.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400015174.mm.1 NM_001170981.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400015174.mm.1 NM_001170982.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400015174.mm.1 NM_001170982.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400015174.mm.1 NM_001170983.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400015174.mm.1 NM_001170983.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400015174.mm.1 NM_001170984.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400015174.mm.1 NM_001170984.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400015174.mm.1 NM_016884.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400015174.mm.1 NM_016884.3 Refseq
Affymetrix Mouse Clariom S Array PSR1400015174.mm.1 TC1400002023.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400015176.mm.1 NM_001170981.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400015176.mm.1 NM_001170981.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400015176.mm.1 NM_001170982.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400015176.mm.1 NM_001170982.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400015176.mm.1 NM_001170983.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400015176.mm.1 NM_001170983.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400015176.mm.1 NM_001170984.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400015176.mm.1 NM_001170984.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400015176.mm.1 NM_016884.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400015176.mm.1 NM_016884.3 Refseq
Affymetrix Mouse Clariom S Array PSR1400015176.mm.1 TC1400002023.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0