This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04206646_g1
See other TCEA1 GE Assays ›
Gene Symbol:
Tcea1
Gene Name:
transcription elongation factor A (SII) 1
Gene Aliases:
S-II
Chromosome Location:
Chr.1: 4857694 - 4897909 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04206646_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tcea1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001159751.1 NP_001153223.1 4 - 5 435 90
NM_011541.4 NP_035671.1 4 - 5 542 90
GenBank mRNA AK131928.1 - 4 - 6 726 90
AK136035.1 - 4 - 5 435 90
AK143593.1 - 4 - 5 400 90
BC006022.1 - 4 - 5 417 3590733 90
BC061490.1 - 4 - 5 434 2582185 90
BC083127.1 - 4 - 5 405 6390364 90
M18209.1 - 3 - 4 338 90
M18210.1 - 4 - 5 396 90

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tcea1

Panther Classification:

Molecular Function -

general transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
erythrocyte differentiation
regulation of DNA-templated transcription, elongation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
nucleic acid binding
DNA binding
protein binding
zinc ion binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100000060.mm.1 NM_001159750.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100000060.mm.1 NM_001159750.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100000060.mm.1 NM_001159751.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100000060.mm.1 NM_001159751.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100000060.mm.1 NM_011541.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100000060.mm.1 NM_011541.4 Refseq
Affymetrix Mouse Clariom S Array PSR0100000060.mm.1 TC0100003861.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0