This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04203795_g1
See other MIA GE Assays ›
Gene Symbol:
Mia
Gene Name:
melanoma inhibitory activity
Gene Aliases:
CD-RAP, Cdrap, Mia1
Chromosome Location:
Chr.7: 27179741 - 27181149 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04203795_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mia
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_019394.3 NP_062267.2 2 - 3 361 94
GenBank mRNA AW047492.1 - 2 - 3 94
BE864464.1 - 2 - 3 361 94
CK794645.1 - 2 - 3 355 94
X94322.1 - 2 - 3 366 94

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mia

Gene Ontology Categories:

Function(s) Process(es)

cell-matrix adhesion
extracellular matrix organization
growth factor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0