This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03992567_s1
See other RPL35A GE Assays ›
Gene Symbol:
Rpl35a
Gene Name:
ribosomal protein L35A
Gene Aliases:
2810431L15Rik, Rpl35
Chromosome Location:
Chr.16: 33056453 - 33060189 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03992567_s1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rpl35a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001130484.1 NP_001123956.1 5 445 85
NM_001130485.1 NP_001123957.1 5 429 85
NM_021338.3 NP_067313.2 5 475 85
GenBank mRNA BB611247.1 - 5 397 85

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1600000460.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rpl35a

Panther Classification:

Molecular Function -

ribosomal protein

Gene Ontology Categories:

Function(s) Process(es)

rRNA processing
translation
ribosomal large subunit biogenesis
structural constituent of ribosome
poly(A) RNA binding

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1997895 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1970087 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 483469 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 12499 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P145664 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1997895 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1970087 - -
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P509107 CJ043213.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P509107 BU530423.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P509107 AV486753.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P509107 BB611247.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P509107 BC027223.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P288669 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30656007 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30656015 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30656000 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30656004 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30656009 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30656013 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30656010 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30656005 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30656019 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30656006 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30656018 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30655999 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P145664 - -
Whole Mouse Genome G4122A A_52_P369054 AK160963.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P509107 CJ043213.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P509107 BU530423.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P509107 AV486753.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P509107 BB611247.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P509107 BC027223.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P288669 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P128001 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P103963 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P107188 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126137 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116582 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P145664 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12878166 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30656010 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30656006 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30656038 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30656050 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30655994 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30655998 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30656078 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30656033 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30656037 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30656071 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30656016 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30656064 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30656054 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30656060 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30656035 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30656027 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30656004 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30655997 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30656000 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30656002 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30656019 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30655995 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30656055 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30656009 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30656021 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30656052 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30656007 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30656048 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30656065 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30656012 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30656018 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30656070 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30656056 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30656073 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30656042 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30656024 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30656076 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30656032 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30656061 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30656005 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30656066 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30655999 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30656068 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30656023 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30656030 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30655990 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30656083 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30656082 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30656046 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30656059 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30656044 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30656009 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30656019 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30656007 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30656010 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30656000 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30656012 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12878166 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12878158 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12878161 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12956609 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12652344 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12969574 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30656005 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30656002 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30656013 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30656004 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10407508 - -
Affymetrix Mouse Gene 1.0 ST Array 10489189 - -
Affymetrix Mouse Gene 1.0 ST Array 10435189 - -
Affymetrix Mouse Gene 1.0 ST Array 10533627 - -
Affymetrix Mouse Gene 1.0 ST Array 10437628 - -
Affymetrix Mouse Gene 1.0 ST Array 10521037 - -
Affymetrix Mouse Gene 1.0 ST Array 10360740 - -
Affymetrix Mouse Gene 1.0 ST Array 10544541 - -
Affymetrix Mouse Gene 1.0 ST Array 10485686 - -
Affymetrix Mouse Gene 1.0 ST Array 10489187 - -
Affymetrix Mouse Gene 1.0 ST Array 10489188 - -
Affymetrix Mouse Gene 1.0 ST Array 10435190 - -
Affymetrix Mouse Gene 1.0 ST Array 10360738 - -
Affymetrix Mouse Gene 1.0 ST Array 10360739 - -
Affymetrix Mouse Gene 1.0 ST Array 10435192 - -
Affymetrix Mouse Gene 1.0 ST Array 10449550 - -
Affymetrix Mouse Gene 1.0 ST Array 10435191 - -
Affymetrix Mouse Gene 1.0 ST Array 10538081 - -
Affymetrix Mouse Gene 1.0 ST Array 10435456 - -
Affymetrix Mouse Gene 1.0 ST Array 10435188 - -
Affymetrix Mouse Gene 1.1 ST Array 10544540 - -
Affymetrix Mouse Gene 1.1 ST Array 10360737 - -
Affymetrix Mouse Gene 1.1 ST Array 10538080 - -
Affymetrix Mouse Gene 1.1 ST Array 10437627 - -
Affymetrix Mouse Gene 1.1 ST Array 10435455 - -
Affymetrix Mouse Gene 1.1 ST Array 10489186 - -
Affymetrix Mouse Gene 1.1 ST Array 10533626 - -
Affymetrix Mouse Gene 1.1 ST Array 10435187 - -
Affymetrix Mouse Gene 1.1 ST Array 10449549 - -
Affymetrix Mouse Gene 1.1 ST Array 10485685 - -
Affymetrix Mouse Gene 1.1 ST Array 10521036 - -
Affymetrix Mouse Gene 1.1 ST Array 10407507 - -
Mouse Genome 430 2.0 Array 1417317_s_at - -
Mouse Genome 430 2.0 Array 1417317_PM_s_at - -
Affymetrix Mouse Clariom S Array PSR1600004359.mm.1 NM_001130484.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600004359.mm.1 NM_001130484.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600004359.mm.1 NM_001130485.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600004359.mm.1 NM_001130485.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600004359.mm.1 NM_021338.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600004359.mm.1 NM_021338.3 Refseq
Affymetrix Mouse Clariom S Array PSR1600004359.mm.1 TC1600000460.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600004357.mm.1 NM_001130484.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600004357.mm.1 NM_001130484.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600004357.mm.1 NM_001130485.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600004357.mm.1 NM_001130485.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600004357.mm.1 NM_021338.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600004357.mm.1 NM_021338.3 Refseq
Affymetrix Mouse Clariom S Array PSR1600004357.mm.1 TC1600000460.mm.1 MTA TC
Mouse Expression Set 430 1417317_s_at - -
Mouse Expression Set 430 1417317_PM_s_at - -
Murine Genome U74v2 Set 93513_at - -
Murine Genome U74v2 Set 93513_PM_at - -
Mouse Genome 430A 2.0 Array 1417317_s_at - -
Mouse Genome 430A 2.0 Array 1417317_PM_s_at - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 1850541 - -
Illumina mouse-6 v1.0 expression beadchip 1850541 - -
Illumina mouseRef-8 v1.0 expression beadchip 0001850541 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2595967 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2595967 - -
mouse-6 v1.1 (Illumina) scl057808.2_82-S - -
Illumina Sentrix mouse-6.v1 bead array 1850541 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl057808.2_82-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 1850541 - -
MouseRef-8 v2 Expression BeadChip ILMN_2595967 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2595967 - -
Illumina Sentrix Mouse-6 Expression BeadChip (3) scl057808.2_82-S - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0