This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03991041_g1
See other ARFGAP2 GE Assays ›
Gene Symbol:
Arfgap2
Gene Name:
ADP-ribosylation factor GTPase activating protein 2
Gene Aliases:
2310032E02Rik, Zfp289
Chromosome Location:
Chr.2: 91265115 - 91277371 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03991041_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Arfgap2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001166024.1 NP_001159496.1 3 - 4 484 87
NM_023854.2 NP_076343.2 3 - 4 484 87
GenBank mRNA AF229439.1 - 3 - 4 293 87
AK009565.1 - 3 - 4 308 87
AK149837.1 - 3 - 4 319 87
AK168489.1 - 3 - 4 294 87
AK169144.1 - 3 - 4 307 87
BC005495.1 - 3 - 4 332 3487811 87

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Arfgap2

Panther Classification:

Molecular Function -

GTPase-activating protein

Gene Ontology Categories:

Function(s) Process(es)

transport
protein transport
vesicle-mediated transport
GTPase activator activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0