This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03980138_g1
See other FAM155A GE Assays ›
Gene Symbol:
Fam155a
Gene Name:
family with sequence similarity 155, member A
Gene Aliases:
6430500D16, AW121567, Tmem28
Chromosome Location:
Chr.8: 9206010 - 9773363 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03980138_g1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Fam155a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK149226.1 - 2 - 3 260 100
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY742096.1 - 2 - 3 260 Chr. 8 - 9430007 - 9770104 [-] on Build GRCm38 100

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream RIKEN cDNA 4930453L07 gene 4930453L07Rik -425.2kb Anti-sense Chr. 8 - 9144669 - 9149050 on NCBI Build GRCm38
Downstream ligase IV,
DNA,
ATP-dependent
Lig4 5830471N16Rik,
tiny
395.2kb Sense Chr. 8 - 9970020 - 9976323 on NCBI Build GRCm38

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0