This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03975435_m1
See other LOC102640140 GE Assays ›
Gene Symbol:
LOC102640140
Gene Name:
uncharacterized LOC102640140
Gene Aliases:
-
Chromosome Location:
Chr.9: 42031922 - 42037042 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03975435_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC102640140
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK037519.1 - 617 71
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BB630713.1 - 3 - 4 613 Chr. 9 - 42022617 - 42033382 [+] on Build GRCm38 71

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream sortilin-related receptor,
LDLR class A repeats-containing
Sorl1 2900010L19Rik,
AI596264,
AW261561,
LR11,
SorLA,
gp250,
mSorLA
Anti-sense Chr. 9 - 41968489 - 42124289 on NCBI Build GRCm38
Downstream predicted gene,
39321
Gm39321 108.7kb Sense Chr. 9 - 42142092 - 42145394 on NCBI Build GRCm38

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Mus musculus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0